Skip to Content
Merck
All Photos(2)

Key Documents

1.46081

Millipore

Sabouraud Dextrose Agar

Tween®, Lecithin, lid, ICR: non-lockable, plate diam. 90 mm, sterile; irradiated, triple packed of 120 plates, suitable for air monitoring

Synonym(s):

ICR plate, SDA, SDA with neutralizers

Sign Into View Organizational & Contract Pricing

Select a Size

120 PKG
€404.00

€404.00


Available to ship on31 March 2025Details


Request more information

Select a Size

Change View
120 PKG
€404.00

About This Item

UNSPSC Code:
41106227
NACRES:
NA.74

€404.00


Available to ship on31 March 2025Details


Request more information

material

polystyrene plate

Quality Level

Agency

EP
USP

sterility

sterile; irradiated

form

plate (ready-to-use)

feature

lid ICR: non-lockable
ready-to-use

composition

Lecithin
Tween®

packaging

triple packed of 120 plates

plate diam.

90 mm

pH

5.6 ( in H2O)

application(s)

air monitoring
pharmaceutical

compatibility

for use with MAS-100®

storage temp.

2-25°C

suitability

molds
yeasts

General description

Sabouraud Dextrose Agar + LT- ICR in 90 mm settle plates (non-lockable) is designed for the determination of the total count of yeasts and molds in air via active or passive air monitoring as well as fingerprints of personnel in isolators and clean rooms. Sabouraud Dextrose Medium contains digests of animal tissues (peptones) which provide a nutritious source of amino acids and nitrogenous compounds for the growth of fungi and yeasts. Dextrose is added as the energy and carbon source. The low pH favours fungal growth. Lecithin and polysorbate 80 (Tween®) are neutralizers to inactivate the active agents of disinfectants such as Alcohol (70% ethanol or isopropyl alcohol), Dichloroisocyanurate, Glucoprotamine, Peracetic, quaternary ammonium compounds and Biguanides. To differentiate Tryptic Soy Agar (TSA) from Sabouraud Dextrose agar (SDA), SDA ICR media are filled in pink colored dish.

Application

Sabouraud Dextrose Agar (SDA) is a complex medium for cultivation and isolation of yeasts and molds. The media finds its application in environmental monitoring by determining the yeast and mold count in air through active or passive air monitoring as well as fingerprints of personnel in isolators and clean rooms.

Legal Information

MAS-100 is a registered trademark of MBV AG
TWEEN is a registered trademark of Croda International PLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Microbial monitoring; environmental monitoring; interim storage; ICR contact plates; contact plates; TSA w. LTHThio Contact ICR+

Related Content

Settle plates for isolators and critical cleanrooms

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service