Skip to Content
Merck
All Photos(1)

Key Documents

14-605

Sigma-Aldrich

Fer Protein, active, 10 µg

Sign Into View Organizational & Contract Pricing

Select a Size

14-605
€468.00

€468.00


Estimated to ship on04 April 2025Details


Request a Bulk Order

Select a Size

Change View
14-605
€468.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405

€468.00


Estimated to ship on04 April 2025Details


Request a Bulk Order

recombinant

expressed in baculovirus infected Sf21 cells

Quality Level

description

protein target Fer

purified by

(Ni2+/NTA-agarose)

technique(s)

activity assay: suitable

UniProt accession no.

Gene Information

human ... FER(2241)

General description

FUNCTION: SwissProt: P16591 # Tyrosine kinase of the non-receptor type. Probably performs an important function, perhaps in regulatory processes such as cell cycle control.
SIZE: 822 amino acids; 94638 Da
SUBCELLULAR LOCATION: Cytoplasm. Nucleus. Note=Associated with the chromatin.
TISSUE SPECIFICITY: Expressed in a variety of lymphoid cell lines.
SIMILARITY: SwissProt: P16591 ## Belongs to the protein kinase superfamily. Tyr protein kinase family. Fes/fps subfamily. & Contains 1 FCH domain. & Contains 1 protein kinase domain. & Contains 1 SH2 domain.

Quality

routinely evaluated by phosphorylation of IGF-IRtide (Catalog #12-527)

Physical properties

35.6kDa

Storage and Stability

6 months at -70°C

Analysis Note

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service