Skip to Content
Merck
All Photos(1)

Key Documents

14-512-M

Sigma-Aldrich

Axl Protein, active, 10 µg

Active, N-terminal His6-tagged, recombinant amino acids 473-end human Axl, for use in Kinase Assays.

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€578.00
250 μG
€3,770.00

€578.00


Estimated to ship on25 April 2025Details



Select a Size

Change View
10 μG
€578.00
250 μG
€3,770.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

€578.00


Estimated to ship on25 April 2025Details


biological source

human

Quality Level

recombinant

expressed in Sf21 cells

mol wt

Mw 48 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

Gene Information

human ... AXL(558)

General description

N-terminal His6-tagged, recombinant amino acids 473-end human Axl
Product Source: expressed in Sf21 cells

Application

Research Category
Metabolism
Research Sub Category
Obesity

Metabolic Disorders

Biochem/physiol Actions

Protein Target: Axl
Target Sub-Family: TK

Packaging

Also available in 250 µg size --call for pricing and availability and reference catalog number 14-512M when ordering the 250 µg size.

Quality

routinely evaluated by phosphorylation of Axl-tide

Physical form

Ni2+/NTA-agarose

Storage and Stability

6 months at -70°C

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service