Skip to Content
Merck
All Photos(1)

Key Documents

14-500-M

Sigma-Aldrich

Flt-3 Protein, active, 10 µg

Active, N-terminal GST tagged, recombinant human Flt3, amino acids 564 -end. For use in Kinase Assays.

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€547.00
250 μG
€3,470.00

€547.00


Estimated to ship on24 April 2025Details


Request a Bulk Order

Select a Size

Change View
10 μG
€547.00
250 μG
€3,470.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41

€547.00


Estimated to ship on24 April 2025Details


Request a Bulk Order

biological source

human

Quality Level

recombinant

expressed in baculovirus infected Sf21 cells

mol wt

Mw 77.4 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

Gene Information

human ... FLT3(2322)

General description

N-terminal GST tagged, recombinant human Flt3, amino acids 564 - end
Product Source: expressed in Sf21 cells

Application

Active, N-terminal GST tagged, recombinant human Flt3, amino acids 564 -end, for use in Kinase Assays.

Biochem/physiol Actions

Protein Target: Flt3
Target Sub-Family: TK

Packaging

Also available in 250μg size (2x125μg)--call for pricing and availability and reference catalog number 14-500M when ordering the 250μg size.

Quality

routinely evaluated by phosphorylation of Abltide

Physical form

Glutathione Sepharose

Storage and Stability

6 months at -70°C

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

Sepharose is a trademark of Cytiva
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service