Skip to Content
Merck
All Photos(1)

Key Documents

14-451-M

Sigma-Aldrich

ROKα/ROCK-II Protein, active, 10 µg

Active, N-terminal His6-tagged recombinant human ROCK-II residues 11-552, for use in Kinase Assays.

Synonym(s):

ROK, Renal carcinoma antigen NY-REN-35, Rho kinase, Rho-associated, coiled-coil-containing protein kinase 1, p160ROCK

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€547.00
250 μG
€3,470.00

€547.00


Estimated to ship on25 April 2025Details


Request a Bulk Order

Select a Size

Change View
10 μG
€547.00
250 μG
€3,470.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41

€547.00


Estimated to ship on25 April 2025Details


Request a Bulk Order

biological source

human

Quality Level

recombinant

expressed in baculovirus infected Sf21 cells

form

liquid

shelf life

6 mo.

mol wt

Mw 63.3 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

solubility

soluble

NCBI accession no.

UniProt accession no.

storage temp.

−70°C

Gene Information

human ... ROCK1(6093)

General description

Research area: APOPTOSIS

N-terminal His6-tagged recombinant human ROCK-II residues 11-552
Product Source: expressed by baculovirus in Sf21 insect cells

Rho kinase (ROCK) is a type of serine/threonine kinase. There are two isoforms, ROCK1 and ROCK2. In humans ROCK1 and ROCK2 are composed of 33 exons and located on chromosome 18 (18q11.1). [1] The ROCK protein structure is characterized by a catalytic kinase domain and Rho-binding domain (RBD) at N-terminal and pleckstrin-homology (PH) domain and an internal cysteine-rich (CR) domain at C-terminal.

Biochem/physiol Actions

Protein Target: ROKα/ROCK-II
Target Sub-Family: AGC

ROCK (Rho kinase) is activated by interacting with Rho GTPase enzyme. ROCK plays a crucial role in various essential cellular functions like adhesion, contraction, migration, and proliferation. Moreover, it regulates the cytoskeleton and actomyosin contractility during apoptosis. Overexpression of ROCK protein has been observed in various types of cancer such as osteosarcoma, hepatocellular carcinomas, colon and bladder cancers.[2]

Packaging

Also available in 250μg size (14-451M), please inquire for pricing and availability.

Quality

routinely evaluated by phosphorylation of the Long S6 Kinase substrate peptide

Physical form

Ni2+/NTA-agarose

Storage and Stability

6 months at -70°C

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Sahai et al.
Current biology : CB, 9(3), 136-145 (1999-02-18)
The Ras-related GTPase RhoA controls signalling processes required for cytoskeletal reorganisation, transcriptional regulation, and transformation. The ability of RhoA mutants to transform cells correlates not with transcription but with their ability to bind ROCK-I, an effector kinase involved in cytoskeletal
J Feng et al.
The Journal of biological chemistry, 274(6), 3744-3752 (1999-01-28)
Rho-associated kinase (Rho-kinase) from chicken gizzard smooth muscle was purified to apparent homogeneity (160 kDa on SDS-polyacrylamide gel electrophoresis) and identified as the ROKalpha isoform. Several substrates were phosphorylated. Rates with myosin phosphatase target subunit 1 (MYPT1), myosin, and the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service