Skip to Content
Merck
All Photos(1)

Documents

14-415

Sigma-Aldrich

Ral Assay Reagent (Ral BP1, agarose)

Synonym(s):

Agarose Ral Assay, Ral BP1 Assay Reagent, Ral Detection Reagent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
eCl@ss:
32160405
NACRES:
NA.41

form

liquid

Quality Level

manufacturer/tradename

Upstate®

technique(s)

affinity binding assay: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

General description

GST fusion-protein, corresponding to residues 397-519 of rat Ral binding protein (RalBP1) bound to agarose. Also referred to as RLIP and RLP; specifically binds to and precipitates RalA-GTP and RalB-GTP from cell lysates.
Product Source: Expressed in E. coli.

Quality

Routinely evaluated by Affinity Precipitation/Immunoblot Analysis

Physical form

300 μg of protein in 600 μL of a 50% agarose slurry in 50 mM Tris, pH 7.5, 200 mM NaCl, 1% Triton X-100, 20 μM PMSF, 1 mM aprotinin prior to the addition of glycerol to 50%. Liquid suspension.

Storage and Stability

1 year at -20°C

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

EU REACH Annex XVII (Restriction List)

EU REACH Annex XIV (Authorisation List)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Goi et al.
Molecular and cellular biology, 19(3), 1731-1741 (1999-02-18)
Ras proteins can activate at least three classes of downstream target proteins: Raf kinases, phosphatidylinositol-3 phosphate (PI3) kinase, and Ral-specific guanine nucleotide exchange factors (Ral-GEFs). In NIH 3T3 cells, activated Ral-GEFs contribute to Ras-induced cell proliferation and oncogenic transformation by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service