Skip to Content
Merck
All Photos(1)

Key Documents

1.44021

Millipore

Hycon® Contact Slides

D/E (Dey/Engley) Neutralizing Agar, Polysorbate 80, for bacteria, for fungi, γ-irradiated, double packed of 20 units, suitable for surface monitoring

Synonym(s):

Hycon® Contact Slides DE-y, Neutralizing Agar

Sign Into View Organizational & Contract Pricing

Select a Size

20 UNITS
€91.30

€91.30


Estimated to ship on31 March 2025Details


Request more information

Select a Size

Change View
20 UNITS
€91.30

About This Item

UNSPSC Code:
41106200
NACRES:
NA.74

€91.30


Estimated to ship on31 March 2025Details


Request more information

Quality Level

sterility

γ-irradiated

form

plate (ready-to-use)

feature

ready-to-use

composition

Polysorbate 80

packaging

double packed of 20 units

manufacturer/tradename

HYCON®

pH

7.1 ( in H2O)

mp

>95 °C

application(s)

pharmaceutical
surface monitoring

compatibility

stand alone

storage temp.

2-8°C

suitability

bacteria
fungi

General description

Contact Slides DE-y are developed for easy and flexible microbiological monitoring of flat and curved surfaces. The unique design makes it a universal tool for aseptic environments and isolators. Each contact slide is individually sealed and irradiated D/E (Dey/Engley) Neutralizing Agar is a complex medium for cultivation and isolation of a wide range of bacteria, yeasts and molds. The medium is supplemented with a specific mixture of neutralizers developed to neutralize a wide variety of antimicrobial active agents at the sampling point such as phenols and quaternary ammonium compounds

Application

Contact Slides DE-y are designed for the determination of the total microbial count on dry, sanitized surfaces as well as personnel in controlled clean room environments of production lines, i.e. grade A and B cleanrooms, RABS or isolators.

Legal Information

HYCON is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

For easy and flexible microbiological monitoring of surfaces in the food & beverage industry.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service