Skip to Content
Merck
All Photos(1)

Documents

126870

Sigma-Aldrich

Alsterpaullone

A cell-permeable, potent, reversible, and ATP competitive inhibitor of GSK-3β (IC₅₀ = 4 nM) and Cdk1/cyclin B (IC₅₀ = 35 nM).

Synonym(s):

Alsterpaullone, 9-Nitro-7,12-dihydroindolo[3,2-d][1]benzazepin-6(5H)-one, 9-Nitropaullone, NSC 705701

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H11N3O3
CAS Number:
Molecular Weight:
293.28
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light yellow to brown

solubility

DMSO: 20 mg/mL

shipped in

ambient

storage temp.

−20°C

InChI

1S/C16H11N3O3/c20-15-8-12-11-7-9(19(21)22)5-6-14(11)18-16(12)10-3-1-2-4-13(10)17-15/h1-7,18H,8H2,(H,17,20)

InChI key

OLUKILHGKRVDCT-UHFFFAOYSA-N

General description

A cell-permeable, potent, reversible, and ATP competitive inhibitor of GSK-3β (IC50 = 4 nM) and Cdk1/cyclin B (IC50 = 35 nM). Displays remarkable in vitro antitumor activity. Inhibits Tau phosphorylation at sites that are typically phosphorylated by GSK-3β in Alzheimer’s disease. Also inhibits Cdk5/p25-dependent phosphorylation of DARPP-32.
A cell-permeable, potent, reversible, and ATP competitive inhibitor of cdk1/cyclin B (IC50 = 35 nM) and GSK-3β (IC50 = 4 nM). One of the most active paullones that acts by competing with ATP for binding to GSK-3β and inhibits the phosphorylation of tau. Also inhibits the activity of Cdk5/p25-dependent phosphorylation of DARPP-32 (a 32 kDa dopamine and adenosine 3′,5′-monophosphate-regulated phosphoprotein). Displays remarkable antitumor activity in vitro. Inhibits growth of the colon cancer cell line HCT-116, with an IC50 in the nanomolar range.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
GSK-3β, Cdk1/cyclin B
Product competes with ATP.
Reversible: yes
Target IC50: 4 nM, 35 nM, against GSK-3β, Cdk1/cyclin B, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Leost, M., et al. 2000. Eur. J. Biochem.267, 5983.
Schultz, C., et al. 1999. J. Med. Chem. 42, 2909.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marissa Cloutier et al.
Nature communications, 13(1), 2516-2516 (2022-05-07)
X-chromosome inactivation is a paradigm of epigenetic transcriptional regulation. Female human embryonic stem cells (hESCs) often undergo erosion of X-inactivation upon prolonged culture. Here, we investigate the sources of X-inactivation instability by deriving new primed pluripotent hESC lines. We find

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service