manufacturer/tradename
Upstate®
technique(s)
activity assay: suitable (kinase)
NCBI accession no.
UniProt accession no.
shipped in
dry ice
Gene Information
human ... PRKCA(5578)
Biochem/physiol Actions
Protein Target: PKC
Target Sub-Family: AGC
Quality
Routinely evaluated as a substrate for PKC Epsilon (14-236), PKC Zeta (14-235) and MST2 activity (14-417).
Physical form
Lyophilized powder
Storage and Stability
Lyophilized: 2 years at -20°C; Rehydrated: 6 months at -20°C.
Legal Information
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Expression, purification, and characterization of protein kinase C-epsilon
Schaap, D. and Parker, P. J.
The Journal of Biological Chemistry, 265, 7301-7307 (1990)
D Schaap et al.
FEBS letters, 243(2), 351-357 (1989-01-30)
PKC-epsilon was isolated from a murine brain cDNA library. The clone, lambda 61PKC-epsilon, encoded a polypeptide of 737 amino acids that is homologous to other PKCs. Northern analysis showed that the 7 kb mRNA for this cDNA is widely expressed.
H L Lai et al.
The Journal of biological chemistry, 272(8), 4970-4977 (1997-02-21)
We have previously reported that phosphorylation of adenylyl cyclase type VI (AC6) may result in the suppression of adenylyl cyclase activity during desensitization of the A2a-adenosine receptor-mediated cAMP response (A2a desensitization) in rat pheochromocytoma PC12 cells. In the present study
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service