Skip to Content
Merck
All Photos(1)

Key Documents

124031

Sigma-Aldrich

InhibitorSelect Akt/PI 3-K/mTOR Signaling Pathway Inhibitor Panel

The InhibitorSelect Akt/PI 3-K/mTOR Signaling Pathway Inhibitor Panel controls the biological activity of InhibitorSelect.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

shipped in

ambient

storage temp.

−20°C

General description

A panel of 12 highly potent and selective kinase inhibitors and a negative control useful for the study of Akt/PI 3-K/mTOR signaling pathway. This panel contains the following inhibitors: 1 mg of Akt Inhibitor IV (Cat. No. 124011); 1 mg of Akt Inhibitor VIII, Isozyme-Selective, Akti-1/2 (Cat. No.124018); 5 mg of LY-294002 (Cat. No. 440202); 1 mg of LY 303511 (Cat. No. 440203); 5 mg of PDK1/Akt/Flt Dual Pathway Inhibitor (Cat. No. 521275); 1 mg of PI-103 (Cat. No. 528100); 5 mg of PI 3-Kγ Inhibitor (Cat. No. 528106); 5 mg of PI 3-Kγ Inhibitor II (Cat. No. 528108); 5 mg of PI 3-Kα Inhibitor IV (Cat. No. 528111); 5 mg of PI 3-Kα Inhibitor VIII (Cat. No. 528116); 100 µg of Rapamycin (Cat. No. 553210); 500 µg of Ro-31-8220 (Cat. No. 557520); and 1 mg of Wortmannin (Cat. No. 681675). Also provided is 15 ml of anhydrous DMSO. Supplied with a data sheet.

Packaging

Packaged under inert gas

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS (O)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C) each inhibitor. Each inhibitor solution is stable for up to 3 months at -20°C.
Please refer to information provided on the CD for reconstitution information of individual inhibitors.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service