Skip to Content
Merck
All Photos(1)

Key Documents

124015

Sigma-Aldrich

Akt Inhibitor IV

InSolution, ≥98%

Synonym(s):

InSolution Akt Inhibitor IV

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€163.00

€163.00


Estimated to ship on14 April 2025Details



Select a Size

Change View
1 MG
€163.00

About This Item

Empirical Formula (Hill Notation):
C31H27IN4S
Molecular Weight:
614.54
UNSPSC Code:
12352200
NACRES:
NA.77

€163.00


Estimated to ship on14 April 2025Details


Quality Level

Assay

≥98% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

shipped in

wet ice

storage temp.

2-8°C

General description

A cell-permeable benzimidazole compound that inhibits Akt phosphorylation/activation presumably by targeting the ATP binding site of a kinase upstream of Akt, but downstream of PI3K. Shown to block Akt-mediated FOXO1a nuclear export (IC50 = 625 nM) and cell proliferation (IC50<1.25 µM) in 786-O cells. Unlike phosphatidylinositol analog-based Akt inhibitors (Cat. Nos. 124005, 124008, 124009), this inhibitor does not affect PI3K.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Akt
Product does not compete with ATP.
Reversible: yes
Target IC50: 625 nM and <1.25 µM in blocking Akt-mediated FOXO1a nuclear export and cell proliferation, respectively, in 786-O cells

Packaging

Packaged under inert gas
Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Irritant (B)

Physical form

A 10 mM (1 mg/163 µl) solution of Akt Inhibitor IV (Cat. No. 124011) in DMSO.

Reconstitution

Following initial use, aliquot and refrigerate (4°C).

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - closed cup - (Dimethylsulfoxide)

Flash Point(C)

87 °C - closed cup - (Dimethylsulfoxide)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seong-Hoon Yun et al.
Anticancer research, 40(1), 177-190 (2020-01-02)
The chicken ovalbumin upstream promoter-transcription factor II (COUP-TFII) regulates cancer cell proliferation and invasion via complex molecular mechanisms. We aimed to investigate whether COUP-TFII modulates proliferation and invasion of the colorectal adenocarcinoma cell line HT-29. HT-29 cells were stably tranfected

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service