Skip to Content
Merck
All Photos(1)

Key Documents

12-385

Sigma-Aldrich

Crosstide, biotin conjugate

Crosstide, biotin conjugate primarily used in Kinase Assays.

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
€244.00

€244.00


Estimated to ship on23 April 2025Details



Select a Size

Change View
500 μG
€244.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.42

€244.00


Estimated to ship on23 April 2025Details


Quality Level

form

liquid

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

shipped in

dry ice

Biochem/physiol Actions

Protein Target: MSK1
Target Sub-Family: AGC

Quality

Routinely evaluated as a substrate with serine/threonine kinases.

Physical form

sterile water

Storage and Stability

1 year at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D R Alessi et al.
The EMBO journal, 15(23), 6541-6551 (1996-12-02)
Insulin activated endogenous protein kinase B alpha (also known as RAC/Akt kinase) activity 12-fold in L6 myotubes, while after transfection into 293 cells PKBalpha was activated 20- and 50-fold in response to insulin and IGF-1 respectively. In both cells, the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service