Skip to Content
Merck
All Photos(1)

Key Documents

12-330

Sigma-Aldrich

Casein Kinase 2 Substrate Peptide

Casein Kinase 2 Substrate Peptide primarily used in Kinase Assays, liquid

Sign Into View Organizational & Contract Pricing

Select a Size

1 μMOL
€378.00

€378.00


Estimated to ship on15 April 2025Details



Select a Size

Change View
1 μMOL
€378.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

€378.00


Estimated to ship on15 April 2025Details


Product Name

Casein Kinase 2 Substrate Peptide, Casein Kinase 2 Substrate Peptide primarily used in Kinase Assays.

Quality Level

form

liquid

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

Gene Information

human ... CSNK2B(1460)

Application

Casein Kinase 2 Substrate Peptide primarily used in Kinase Assays.

Biochem/physiol Actions

Protein Target: CK2
Target Sub-Family: CMGC

Quality

Routinely evaluated as a substrate in a kinase assay.

Physical form

20 mM MOPS, pH 7.2; 25 mM b-glycerol phosphate; 5 mM EGTA; 1 mM sodium orthovanadate; 1 mM dithiothreitol

Storage and Stability

Stable for 2 years at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of the product, centrifuge the original vial prior to removing the cap.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service