Skip to Content
Merck
All Photos(1)

Key Documents

12-240

Sigma-Aldrich

MAPKAP Kinase Substrate Peptide

MAPKAP Kinase Substrate Peptide primarily used in Kinase Assays.

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€310.00

€310.00


Estimated to ship on28 April 2025Details



Select a Size

Change View
1 MG
€310.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41

€310.00


Estimated to ship on28 April 2025Details


Quality Level

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

Gene Information

human ... MAPKAPK2(9261)

Biochem/physiol Actions

Protein Target: MAPKAP
Target Sub-Family: CAMK

Quality

Routinely evaluated as a substrate for MAPKAP Kinase 2, active (14-216 & 14-337). May also be used as a substrate for MAPKAP Kinase 1.

Physical form

Lyophilized powder

Storage and Stability

Lyophilized: Stable for 2 years at 4°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Motoneuron apoptosis is blocked by CEP-1347 (KT 7515), a novel inhibitor of the JNK signaling pathway
Maroney, A. C., et al
The Journal of Neuroscience, 18, 104-111 (1998)
M Suomalainen et al.
The EMBO journal, 20(6), 1310-1319 (2001-03-17)
Nuclear targeting of adenovirus is mediated by the microtubule-dependent, minus-end-directed motor complex dynein/dynactin, in competition with plus- end-directed motility. We demonstrate that adenovirus transiently activates two distinct signaling pathways to enhance nuclear targeting. The first pathway activates integrins and cAMP-dependent

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service