Skip to Content
Merck
All Photos(1)

Key Documents

12-110

Sigma-Aldrich

Phosphotyrosine control (EGF-stim A431 cell lysate)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€465.00

€465.00


Estimated to ship on18 February 2025Details



Select a Size

Change View
1 MG
€465.00

About This Item

UNSPSC Code:
41105330
eCl@ss:
32011807
NACRES:
NA.42

€465.00


Estimated to ship on18 February 2025Details


biological source

human

Quality Level

form

liquid

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

shipped in

dry ice

Cell Line Description

A431

Quality

routinely evaluated as a positive control for Western Blot and Immunoprecipitation

Physical form

RIPA buffer

Storage and Stability

6 months at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XIV (Authorisation List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Malgorzata Kwasniak-Owczarek et al.
Methods in molecular biology (Clifton, N.J.), 2363, 183-197 (2021-09-22)
Mitochondria are subcellular organelles with their own genome and expression system, including translation machinery to make proteins. Several independent studies have shown that translation is an essential regulatory step in expression of the plant mitochondrial genome. Thus, the study of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service