Skip to Content
Merck
All Photos(1)

Key Documents

1.18754

Supelco

Chlorine Dioxide Test Kit, colorimetric

0.020-0.55 mg/L (ClO₂), MQuant®

Synonym(s):

Chlorine Dioxide Visual Test Kit

Sign Into View Organizational & Contract Pricing

Select a Size

300 TESTS
€172.00

€172.00


Estimated to ship on07 April 2025Details


Request a Bulk OrderRequest more information

Select a Size

Change View
300 TESTS
€172.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

€172.00


Estimated to ship on07 April 2025Details


Request a Bulk OrderRequest more information

Product Name

Chlorine Dioxide Test, colorimetric, 0.020-0.55 mg/L (ClO2), for use with MCOLORTEST®

product line

MColortest®

Quality Level

specific analyte(s)

chlorine dioxide

measuring range

0.020-0.55 mg/L (ClO2)

compatibility

for use with MCOLORTEST®

detection method

colorimetric

storage temp.

15-25°C

General description

Chlorine dioxide is used for water disinfection and as a bleaching agent in industrial processes. The MQuant® Chlorine Dioxide test (formally MColortest®) can be used for the determination of chlorine dioxide for example in swimming pool water, drinking water and wastewater.

Method
In weakly acidic solution chlorine dioxide reacts with dipropyl-p-phenylenediamine (DPD) to form a red-violet dye. The chlorine dioxide concentration is measured semiquantitatively by visual comparison of the color of the measurement solution with the color fields of a color card.The unique brilliance of the print and their fine color graduation allow precise analyses, even at the lower end of the concentration range. Due to the long test tubes a sensitive measurement range can be covered.
Using the refill pack 1.18757 containing all the reagents, the original test kit can be used for a very long time, making it a particularly economical and ecological system.

This test can be used for
  • Drinking water
  • Disinfectant solutions
  • Boiler and boiler feed water, cooling water

This test is not suited for seawater.
Method: colorimetric, DPD, with color card and comparator block with long test tubes 0.020 - 0.050 - 0.075 - 0.10 - 0.15 - 0.20 - 0.30 - 0.40 - 0.55 mg/l ClO₂ MColortest®

Application


  • Water Treatment and Disinfection: Chlorine dioxide is extensively used in water treatment for its effectiveness in disinfecting drinking water and wastewater. It is an efficient biocide that helps control biofilms and eliminates pathogens. Recent studies focus on the oxidation of natural organic matter in water treatment plants, evaluating the effectiveness of ClO2 in removing contaminants and its impact on water quality and environmental safety (Özdemir, Environment Protection Engineering, 2020).

  • Environmental Monitoring: Chlorine dioxide testing is essential for environmental monitoring to ensure that disinfection by-products (DBPs) do not pose a risk to human health or the environment. Research has highlighted the importance of monitoring ClO2 levels and its by-products in various ecosystems to prevent adverse ecological impacts (Kinani et al., Critical Reviews in Analytical Chemistry, 2022).

  • Food Safety and Sanitation: ClO2 is used in the food industry for sanitizing equipment and surfaces, as well as for washing fruits and vegetables. It is effective in reducing microbial contamination and extending the shelf life of food products. Analytical methods for detecting ClO2 residues in food items are critical for ensuring consumer safety and compliance with regulatory standards (Fabbricino et al., Chemosphere, 2019).

  • Chromatographic and Mass Spectrometric Analysis: Advanced analytical techniques such as chromatography and mass spectrometry are employed to study the speciation and degradation products of chlorine dioxide. These methodologies are crucial for understanding the chemical behavior of ClO2 in different matrices and for developing accurate detection methods (Roumiguières et al., Critical Reviews in Analytical Chemistry, 2023).

  • Cyclic Voltammetry for ClO2 Detection: Cyclic voltammetry is used for the precise determination of free chlorine and chlorine dioxide levels in various water samples, including seawater aquariums. This electrochemical method provides high sensitivity and specificity, making it suitable for monitoring ClO2 in complex aqueous environments (Ko et al., Chinese Journal of Analytical Chemistry, 2022).

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Preparation of a standard solution for Chlorine dioxide acc. to DIN EN ISO 7393

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service