Skip to Content
Merck
All Photos(1)

Key Documents

118605

Sigma-Aldrich

Capping Reagent for DNA Synthesis

Tetrahydrofuran/Acetic anhydride 9/1 (v/v), Novabiochem®

Synonym(s):

Tetrahydrofuran/Acetic anhydride 9/1 (v/v), Novabiochem®

Sign Into View Organizational & Contract Pricing

Select a Size

450 ML
€165.00
4 L
€197.00

€165.00


Estimated to ship on25 August 2025Details


Request a Bulk Order

Select a Size

Change View
450 ML
€165.00
4 L
€197.00

About This Item

UNSPSC Code:
41116006
NACRES:
NA.22

€165.00


Estimated to ship on25 August 2025Details


Request a Bulk Order

Quality Level

product line

Novabiochem®

form

liquid

reaction suitability

reaction type: Acetylations
reagent type: derivatization reagent

manufacturer/tradename

Novabiochem®

shipped in

ambient

storage temp.

2-30°C

General description

Pre-mixed ready-to-use Capping Reagent
  • 118605.0450: Compatible with MerMade DNA Synthesisers. Attaches directly to Instrument (28/405 neck finish)
  • 118605.4000: Compatible with MerMade DNA Synthesisers. Stand alone from Instrument (38mm Neck Finish)

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service