Skip to Content
Merck
All Photos(1)

Documents

1.18336

Supelco

Empty cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47101516
NACRES:
SE.11

material

optical glass vial

technique(s)

photometry: suitable

compatibility

for use with Turbiquant 1500
for use with Turbiquant 3000

detection method

turbidimetric

storage temp.

2-30°C

General description

1500/3000 Turbiquant
These empty celles are suitable for the measurement of turbidity using the instrument types Turbiquant 1500 and 3000. The cells have a diameter of 24 mm, are equipped with screw caps, and are delivered in a seperate bowes to protect them from scratching.

Legal Information

TURBIQUANT is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service