Skip to Content
Merck
All Photos(1)

Key Documents

1.18309

Supelco

Sodium dodecyl sulfate

for ion pair chromatography LiChropur

Synonym(s):

Dodecyl hydrogen sulfate sodium salt, Lauryl sulfate sodium salt, SDS, Dodecyl sodium sulfate

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€667.00
100 G
€3,290.00

€667.00


Estimated to ship on02 May 2025Details


Request a Bulk Order

Select a Size

Change View
25 G
€667.00
100 G
€3,290.00

About This Item

Empirical Formula (Hill Notation):
C12H25NaO4S
CAS Number:
Molecular Weight:
288.38
MDL number:
UNSPSC Code:
12352107
EC Index Number:
205-788-1
NACRES:
NB.21

€667.00


Estimated to ship on02 May 2025Details


Request a Bulk Order

Quality Level

Assay

≥99% (acidimetric)

potency

1427 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

technique(s)

ion pair chromatography: suitable

impurities

≤2% water (Karl Fischer)

transmittance

200 nm, ≥70%
220 nm, ≥90%
250 nm, ≥98%
(0.005 M; 1 cm in H2O)

pH

6.0-7.5 (20 °C, 100 g/L in H2O)

mp

204-207 °C

transition temp

flash point >150 °C

solubility

>130 g/L

density

1.1 g/cm3 at 20 °C

bulk density

490‑560 kg/m3

suitability

passes test for identity (IR)

storage temp.

2-30°C

SMILES string

[Na+].[S](=O)(=O)([O-])OCCCCCCCCCCCC

InChI

1S/C12H26O4S.Na/c1-2-3-4-5-6-7-8-9-10-11-12-16-17(13,14)15;/h2-12H2,1H3,(H,13,14,15);/q;+1/p-1

InChI key

DBMJMQXJHONAFJ-UHFFFAOYSA-M

General description

Sodium dodecyl sulfate (SDS) is commonly used asan ion pair reagent in ion pair chromatography.[1]

Application

Sodium dodecyl sulfate can be used as an ionpair reagent and disperser agent in ion pair-based surfactant-assisteddispersive liquid–liquid micro extraction (IP-SA-DLLME) method for chromiumspeciation analysis.[2]

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 2

Flash Point(F)

338.0 °F

Flash Point(C)

170 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Selective and sensitive speciation analysis of Cr (VI) and Cr (III) in water samples by fiber optic-linear array detection spectrophotometry after ion pair based-surfactant assisted dispersive liquid--liquid microextraction
Yousefi SM and Shemirani F
Journal of Hazardous Materials, 254, 134-140 (2013)
Ion-pair switchable-hydrophilicity solvent-based homogeneous liquid--liquid microextraction for the determination of paraquat in environmental and biological samples before high-performance liquid chromatography
Rahimi K, et al.
Journal of Separation Science, 40(18), 3703-3709 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service