Skip to Content
Merck
All Photos(1)

Documents

1.17985

Supelco

Potassium Test

colorimetric, 250-1500 mg/L (K), MQuant®

Synonym(s):

Potassium chemical test strips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

description

semi-quantitative

Quality Level

product line

MQuant®

specific analyte(s)

potassium

measuring range

250-1500 mg/L (K)

detection method

colorimetric

storage temp.

15-25°C

General description

Method: Colorimetric with test strips and reagent, 250 - 450 - 700 - 1000 - 1500 mg/L Potassium, MQuant®

The MQuant® Potassium test includes test strips and all necessary additional reagents and is used to determine the potassium content of drinking water, mineral water, industrial water and waste water, soil samples, wine, beer and fruit juices. Potassium ions react with dipicrylamine to form an orange-colored complex. The potassium concentration is measured semi-quantitatively by visual comparison of the reaction zone of the test strip with the fields of a color scale. The test strips are perfect for pre-testing a sample in an economic and quick way to estimate the potassium concentration and a possible dilution factor prior to using a quantitative method such as a Reflectoquant® or Spectroquant® Potassium test.

Linkage

Replaces: 117985.0007

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
REFLECTOQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service