Skip to Content
Merck
All Photos(2)

Key Documents

1.17866

Supelco

Glucose Test Strips, colorimetric

greener alternative

10-500 mg/L (glucose), MQuant®

Synonym(s):

Glucose chemical test strips

Sign Into View Organizational & Contract Pricing

Select a Size

50 TESTS
€60.90

€60.90


Available to ship on01 April 2025Details


Request a Bulk OrderRequest more information

Select a Size

Change View
50 TESTS
€60.90

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

€60.90


Available to ship on01 April 2025Details


Request a Bulk OrderRequest more information

Product Name

Glucose Test, colorimetric, 10-500 mg/L (glucose), MQuant®

description

semi-quantitative

Quality Level

product line

MQuant®

analyte chemical class(es)

sugars

reaction suitability

reaction type: enzymatic reaction

greener alternative product characteristics

Waste Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

measuring range

10-500 mg/L (glucose)

detection method

colorimetric

greener alternative category

storage temp.

2-8°C

General description

Method: Colorimetric, with test strips, 10 - 25 - 50 - 100 - 250 - 500 mg/L (glucose), MQuant®

The MQuant® Glucose test is suitable for determining glucose in fruit juices and food. Under the catalytic effect of glucose oxidase, glucose is converted into gluconic acid lactone. In the presence of peroxidase the hydrogen peroxide formed in the process reacts with an organic redox indicator to form a blue-green dye. The glucose concentration is measured semiquantitatively by visual comparison of the reaction zone of the test strip with the fields of a color scale. The test strips are perfect for pre-testing a sample in an economic and quick way to estimate the glucose concentration and a possible dilution factor prior to using a quantitative method such as a Reflectoquant® test.

Design for sustainability benefits:

We continue to demonstrate our commitment to our planet and a sustainable future by exploring ways to improve existing products in our portfolio. We have recently reviewed the current line of MQuant® test strips to identify key opportunities to minimize impact without compromising on the product performance.

This product has benefited from:

  • Test strip with reduction
  • Recycled aluminum tube
  • Digital instruction leaflet
Learn more about the initiative: Small Changes to Test Strips for Big Impact.

Application


  • Bifunctional oxidase-peroxidase mimicking Fe-Ce MOF on paper-based analytical devices to intensify luminol chemiluminescence: This research introduces an innovative glucose assay technique using MOF-enhanced paper devices, providing a cost-effective and accessible method for glucose monitoring in resource-limited settings (Hassanzadeh et al., 2024).

  • Zinc and Silicon Nano-Fertilizers Influence Ionomic and Metabolite Profiles in Maize to Overcome Salt Stress: In this agricultural research, glucose tests are used to monitor the health and metabolic responses of maize to nano-fertilizer treatment, demonstrating the versatility of glucose assays in environmental and plant science applications (Shoukat et al., 2024).

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
REFLECTOQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service