Key Documents
1.17333
Millipore
MAS-100 Atmos® Hardware Key Set (5x)
for use with MAS-100 Atmos®
Select a Size
1173330001
€270.00
€270.00
Estimated to ship on02 June 2025Details
Request more information
Select a Size
Change View
1173330001
€270.00
About This Item
UNSPSC Code:
41103403
NACRES:
NA.74
€270.00
Estimated to ship on02 June 2025Details
Request more information
description
Compressed gas sampling
feature
electronic
manufacturer/tradename
MAS-100 Atmos®
application(s)
compressed gas monitoring
cosmetics
food and beverages
pharmaceutical
compatibility
for use with MAS-100 Atmos®
General description
The MAS-100 Atmos® Hardware Key Set (5x) is designed uniquely for each individual user role. It is available in different colors for easy identification of each user.
The hardware key is a data carrier based on single-wire technology. It contains the login data for accessing the instrument function according to the user management access levels.
Application
The MAS-100 Atmos® Hardware Key Set (5x) is used for access restricted login to operate the instrument.
Features and Benefits
  • 21 CFR part 11 compliant individual login on MAS-100 Atmos®
  • Re-writable
Legal Information
MAS-100 Atmos is a registered trademark of MBV AG