Skip to Content
Merck
All Photos(1)

Key Documents

1.17091

Millipore

Easy clean Base

stainless steel base

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
€3,770.00

€3,770.00


Estimated to ship on20 March 2025Details



Select a Size

Change View
1 PKG
€3,770.00

About This Item

UNSPSC Code:
41122409
NACRES:
NA.74

€3,770.00


Estimated to ship on20 March 2025Details


material

stainless steel base

Agency

non-compendial

feature

installed

packaging

pkg of 1 kit

dilution

(vertical air flow)

application(s)

air monitoring
pharmaceutical

compatibility

for use with MAS-100® Iso

shipped in

ambient

storage temp.

no temp limit

General description

MAS-100 ISO NT® Easy clean Base for Sampling Head is an additional accessory designed to be specifically used for MAS-100 ISO NT® and MAS-100 ISO MH® It is compatible with perforated lids made from stainless steel. The MAS 100 Iso family has been specially developed for monitoring and measuring the microbiological contamination at fixed sampling locations in cleanrooms, RABS and isolators. The sampling head is made of stainless steel with tri-clamps and can be sterilized by gas decontamination or by wiping with cloth. The passive petri dish support is for horizontal mounting only. A tri clamp of 21.7mm internal diameter and 50.5 external diameter is used to connect the tubing with the sampling head base and the instrument. The silicone screen gasket is used to seal the tri clamp connections on instruments of the MAS-100 Iso product family and prevent the instrument from contamination with bigger particles.

Application

MAS-100 ISO NT® Easy clean Base for Sampling Head, Stainless Steel, with Tri-clamp and screen gasket is used for microbial monitoring of ambient air with MAS-100 Iso family microbial air samplers in isolators, RABS and high-grade cleanrooms. Microbial air sampling devices are extensively used in Pharmaceutical applications.

Legal Information

MAS-100 is a registered trademark of MBV AG
MAS-100 ISO MH is a registered trademark of MBV AG
MAS-100 ISO NT is a registered trademark of MBV AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service