1.15946
Nile blue (hydrogen sulfate) (C.I. 51180)
for microscopy Certistain®
Synonym(s):
Nile blue (hydrogen sulfate) (C.I. 51180), Nile blue
About This Item
Recommended Products
Quality Level
form
solid
composition
Dye, ≥90% spectrophotometric assay
pH
2.2 (20 °C, 10 g/L in H2O)
mp
>300 °C
solubility
50 g/L
bulk density
300 kg/m3
εmax
1630-1820 at 635-639 nm in 50% ethanol at 0.003 g/L
application(s)
diagnostic assay manufacturing
hematology
histology
storage temp.
2-30°C
InChI
1S/C20H20N3O.H2O4S/c1-3-23(4-2)13-9-10-17-18(11-13)24-19-12-16(21)14-7-5-6-8-15(14)20(19)22-17;1-5(2,3)4/h5-12H,3-4,21H2,1-2H3;(H2,1,2,3,4)/q+1;/p-1
InChI key
MMYKGGIXVBGGSZ-UHFFFAOYSA-M
General description
As CERTISTAIN dye it is chemically quality-certified according to highest standards and strict specifications. As an IVD product and CE registered, it can be used hassle-free in diagnostics and laboratory accreditations.
Analysis Note
Identity
- Hydrogen sulfate (as sulfur):
passes test
- UV/VIS-Spectrum: passes test
Absorption maximum λ max (ethanol 50 %): 635 - 639 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.003 g/l; ethanol 50 %): 1630 - 1820
TLC-Test: passes test
Loss on drying (110 °C): ≤ 8 %
Suitability for microscopy: passes test
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service