Skip to Content
Merck
All Photos(1)

Documents

1.15940

Sigma-Aldrich

Crystal violet (C.I. 42555)

for microscopy Certistain®

Synonym(s):

Crystal violet (C.I. 42555), Hexamethylenepararosaniline chloride, Gentian violet (C.I. 42555), Hexamethyl-p-rosanilinium, Methyl violet 10 B

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H30ClN3
CAS Number:
Molecular Weight:
407.98
MDL number:
UNSPSC Code:
41116130
EC Index Number:
208-953-6

Quality Level

form

solid

potency

420 mg/kg LD50, oral (Rat)

composition

Dye, ≥88% spectrophotometric assay

pH

2.5-3.5 (20 °C, 10 g/L in H2O)

mp

189-194 °C

solubility

10 g/L

density

1.19 g/cm3 at 20 °C

bulk density

220‑400 kg/m3

εmax

2050-2450 at 589-594 nm in water at 0.002 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-30°C

InChI

1S/C25H30N3.ClH/c1-26(2)22-13-7-19(8-14-22)25(20-9-15-23(16-10-20)27(3)4)21-11-17-24(18-12-21)28(5)6;/h7-18H,1-6H3;1H/q+1;/p-1

InChI key

ZXJXZNDDNMQXFV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Crystal violet (C.I. 42555) for microscopy Certistain®, is a dry dye that can be used to produce a Crystal violet staining solution to differentiate Gram-positive and Gram-negative bacteria in samples of human origin. In the case of Gram-positive bacteria, the bacteria retain their dark blue / dark purple color even after the treatment with decolorizing solution. On the other side Gram-negative bacteria are stained red or pink with the counterstain. The Certistain® dry dye can be used for clinical diagnostic purposes as it is certified and registered as IVD and CE product. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Analysis Note

Dye content (spectrophotometrically): ≥ 88%
Identity (UV/VIS-Spectrum): passes test
Absorption maximum λ max (water): 589 - 594 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.002 g/l; water): 2050 - 2450
TLC-Test: passes test
Michler`s ketone: <0.10%
Loss on drying (110 °C): ≤ 10%
Suitability for microscopy: passes test

Legal Information

CERTISTAIN is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service