Skip to Content
Merck
All Photos(1)

Documents

1.14598

Supelco

Fluoride Test, photometric

photometric, 0.10-20.0 mg/L (F), Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Fluoride Test, photometric, 0.10-20.0 mg/L (F), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 100 tests
sufficient for 250 tests (depending on package size)

specific analyte(s)

fluoride

measuring range

0.10-20.0 mg/L (F)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (Alizarin complexone)

storage temp.

15-25°C

General description

Method: photometric 0.10 - 20.0 mg/l F Spectroquant®
This Spectroquant®Fluoride test allows the accurate quantification of fluoride ions in various waters. Fluoride ions occur on earth in several minerals, but are only present in trace quantities in bodies of water in nature. In a buffered, weakly acidic solution, fluoride ions react with alizarin complexone and lanthanum(III) to form a violet complex that is determined photometrically.
The method is analogous to EPA 340.3 and APHA 4500-F- E.
All our Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.
The Spectroquant® Reagent Test Kits contain highly stable, ready-to-use reagent mixtures to perform the analysis according to the procedure described in the accompanying instruction leaflet.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service