Skip to Content
Merck
All Photos(1)

Key Documents

1.14275

Sigma-Aldrich

Ponceau S (C.I. 27195)

for electrophoresis

Synonym(s):

Ponceau S (C.I. 27195)

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€49.40

€49.40


Estimated to ship on27 March 2025Details



Select a Size

Change View
10 G
€49.40

About This Item

Empirical Formula (Hill Notation):
C22H12N4Na4O13S4
CAS Number:
Molecular Weight:
760.57
MDL number:
UNSPSC Code:
41105305
EC Index Number:
228-319-2
NACRES:
NA.32

€49.40


Estimated to ship on27 March 2025Details


Quality Level

form

solid

pH

9.0 (20 °C, 10 g/L in H2O)

solubility

10 g/L

bulk density

660 kg/m3

storage temp.

2-30°C

InChI

1S/C22H16N4O13S4.4Na/c27-22-20(43(37,38)39)10-12-9-16(41(31,32)33)6-7-17(12)21(22)26-25-18-11-14(3-8-19(18)42(34,35)36)24-23-13-1-4-15(5-2-13)40(28,29)30;;;;/h1-11,25H,(H,28,29,30)(H,31,32,33)(H,34,35,36)(H,37,38,39);;;;/q;4*+1/p-4/b24-23?,26-21-;;;;

InChI key

LZYGEJPERQQCIY-STNZDNLRSA-J

Analysis Note

Identity (UV/VIS-Spectrum): passes test
Absorption maximum λmax. (water): 517 - 523 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.01 g/l; water): 460 - 600
TLC-Test: passes test
Loss on drying (110 °C): ≤ 10 %
Suitability for electrophoresis: passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service