Skip to Content
Merck
All Photos(1)

Key Documents

1.12070

Sigma-Aldrich

Boron

powder

Synonym(s):

Boron

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€66.10
25 G
€103.00
100 G
€345.00
1 KG
€2,210.00

€66.10


Estimated to ship on26 March 2025Details


Request a Bulk Order

Select a Size

Change View
10 G
€66.10
25 G
€103.00
100 G
€345.00
1 KG
€2,210.00

About This Item

Empirical Formula (Hill Notation):
B
CAS Number:
Molecular Weight:
10.81
MDL number:
UNSPSC Code:
12141907
EC Index Number:
231-151-2
NACRES:
NA.22

€66.10


Estimated to ship on26 March 2025Details


Request a Bulk Order

Quality Level

form

powder

autoignition temp.

700 °C

potency

>2000 mg/kg LD50, oral (Rat)

bp

4000 °C/1013 hPa

mp

2075 °C

density

1.73 g/cm3 at 15 °C

bulk density

200‑300 kg/m3

storage temp.

no temp limit

SMILES string

[B]

InChI

1S/B

InChI key

ZOXJGFHDIHLPTG-UHFFFAOYSA-N

Application


  • Ambiphilic boron in 1, 4, 2, 5-diazadiborinine: Examines the unique electronic properties of ambiphilic boron compounds, suggesting potential applications in organic synthesis and materials science (B Wang, Y Li, R Ganguly, H Hirao, R Kinjo, 2016).

  • Metallomimetic chemistry of boron: Discusses boron′s ability to mimic the behaviors of metals in chemical reactions, providing insights into its versatile applications in catalysis and material science (MA Légaré, C Pranckevicius, H Braunschweig, 2019).



Analysis Note

Identity: passes test
Mg (Magnesium): ≤ 1.0 %
Particle Size (d50): about 2 µm

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service