Skip to Content
Merck
All Photos(1)

Key Documents

1.11661

Sigma-Aldrich

Hemacolor® Rapid staining of blood smear

suitable for hematology, staining set for microscopy

Sign Into View Organizational & Contract Pricing

Select a Size

1 SET
€178.00

€178.00


Available to ship on03 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 SET
€178.00

About This Item

UNSPSC Code:
41116121
NACRES:
NA.21

€178.00


Available to ship on03 April 2025Details


Request a Bulk Order

Quality Level

IVD

for in vitro diagnostic use

technique(s)

microbe id | staining: suitable

application(s)

clinical testing
diagnostic assay manufacturing
histology

hematology

storage temp.

15-25°C

General description

The Hemacolor® Rapid staining of blood smear - staining set for microscopy, is used for human-medical cell diagnosis and serves the purpose of the hematological and clinico-cytological investigation of sample material of human origin, for example smears of whole blood and bone marrow.
The Hemacolor® Rapid staining of blood smear staining set combines the brilliance and reproducibility of the Pappenheim staining with the time-advantage of a rapid stain. The Hemacolor® Rapid staining of blood smear staining set contains a fixing solution, one red (eosin) and one blue (azur) staining solution and phosphate buffer tablets pH 7.2 according to Weise (Product number 1.09468). The staining solutions are separately applied, which results in a high degree of stability. The conventionally appearing precipitates in the hematological staining solutions, could be avoided in this kit. In addition to blood and bone marrow smears, Hemacolor® Rapid staining of blood smear can be used in clinical cytology, e.g. for staining urine, sputum, FNAB, effusions and lavages.
A kit contains safe 500-ml glass bottles and six buffer tablets. The package is sufficient for 250 - 1000 applications. The buffer tablets according to Weise pH 7.2 (Product number 1.09468) are also available in larger pack sizes, thus, they can be useful for higher demands. The three solutions needed for staining are all available individually as:

Hemacolor® Rapid staining of blood smear solution 1, fixing solution (Product number 1.11955)
Hemacolor® Rapid staining of blood smear solution 2, colour reagent red (Product number 1.11956)
Hemacolor® Rapid staining of blood smear solution 3, colour reagent blue (Product number 1.11957)

This product is registered as IVD and CE marked. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Analysis Note

Suitability for microscopy (Blood smear): passes test
Erythrocytes: pink to brownish
Nuclei: red-violet
Eosinophilic granules: red to red-brown
Neutrophilic granules: light violet
Lymphocyte cytoplasm: blue

Legal Information

HEMACOLOR is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - Skin Sens. 1 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. I can't find any information of disposal arrangements of Hemacolor 2 and 3. Would you be able to let me know how this product should be disposed of? Many thanks

    1 answer
    1. Hemacolor 2, with catalog number 221024, has its Safety Data Sheet available through an Advanced Search on the website. Disposal information is located in Section 13 of the Safety Data Sheet. For Hemacolor 3, catalog number 221025, the Safety Data Sheet is not available on the website, but a Spanish version was found. The translated disposal information states: "Methods for waste treatment: Waste must be disposed of in accordance with local and national regulations. Do not mix with other waste. Handle dirty containers like the product itself." The Department of Transportation, International Maritime Dangerous Goods, and International Air Transport Association classify it as Not Dangerous Goods.

      Helpful?

  2. Which specific Hemacolor product is suitable for staining bone marrow smears for cytology analysis?

    1 answer
    1. one marrow smears are typically stained with Wright Giemsa or Giemsa, which result in a darker stain with more nuclear detail. Before using Hemacolor reagents for bone marrow smears, it is recommended to compare the results against smears stained with Wright Giemsa or Giemsa.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service