Skip to Content
Merck
All Photos(1)

Key Documents

1.11587

Supelco

Potassium hydroxide solution

c(KOH) = 0.1 mol/l (0.1 N) in methanol, ready-to-use volumetric solution for titration for non-aqueous media, Titriplex®

Synonym(s):

KOH solution in methanol

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€58.50
1 L
€61.40

€58.50


Estimated to ship on27 March 2025Details


Request more information

Select a Size

Change View
1 L
€58.50
1 L
€61.40

About This Item

UNSPSC Code:
12352305
NACRES:
NA.21

€58.50


Estimated to ship on27 March 2025Details


Request more information

Product Name

Potassium hydroxide solution in methanol, c(KOH) = 0.1 mol/l (0.1 N), Titripur®

Quality Level

product line

Titripur®

form

liquid

autoignition temp.

455 °C (Methanol)

quality

Analyzed in our ISO 17025 accredited QC lab

expl. lim.

5.5-44 % (v/v) Methanol)

reaction suitability

reaction type: Acid-base reactions

concentration

0.1 M

technique(s)

titration: suitable

pH

13.5 (20 °C in H2O)

transition temp

flash point 7 °C

density

0.80 g/cm3 at 20 °C

storage temp.

15-25°C

Application


  • Snap-on Adaptor for Microtiter Plates to Enable Continuous-Flow Microfluidic Screening and Harvesting of Crystalline Materials: This study presents an innovative approach for microfluidic screening and harvesting, using a snap-on adapter for microtiter plates, enhancing the crystallization processes in research and development within pharmaceutical and material science applications (Coliaie et al., 2023).

  • A Novel Sample Preparation Method Using Rapid Nonheated Saponification Method for the Determination of Cholesterol in Emulsified Foods: Discusses a novel technique involving potassium hydroxide in methanol for rapidly preparing samples to determine cholesterol levels in food products, beneficial for environmental testing and food safety (Jeong et al., 2012).

  • Extended Release of Bevacizumab by Thermosensitive Biodegradable and Biocompatible Hydrogel: Research focusing on a biodegradable hydrogel for extended drug release, where potassium hydroxide in methanol plays a role in the hydrogel preparation, proving crucial in pharmaceutical applications (Wang et al., 2012).

  • Spectrophotometric and Fluorimetric Determination of Diazepam, Bromazepam, and Clonazepam in Pharmaceutical and Urine Samples: This study utilizes potassium hydroxide in methanol for the enhanced spectrophotometric and fluorimetric analysis of benzodiazepines in clinical and forensic testing scenarios (Salem et al., 2004).

Features and Benefits

Accreditation:
This volumetric solution is analyzed by our calibration laboratory D-K-15185-01-00 which is accredited according to DIN EN ISO/IEC 17025 for analysis of amount-of-substance concentrations in volumetric solutions by DAkkS (Deutsche Akkreditierungsstelle - German National Accreditation Body). The accreditation certificate can be found at www.sigmaaldrich.com/ISO17025.

Packaging

SKU-pack size numbers ending with a "3" are SmartChemicals with an RFID tag on the label for seamless data transfer to the instrument, e.g. xxxxxx1003 or xxxxxxx 4003. More information on www.sigmaaldrich.com/SmartChemicals

Linkage

Replaces: PX1498A-6; PX1498A-3

Analysis Note

Form liquid
Amount-of-substance concentration 0.0995 - 0.1005 mol/L
Measurement uncertainty ± 0.0003 mol/L
Traceability NIST SRM

The concentration is determined by volumetric titration and refers to 20°C.
The amount-of-substance concentration of this volumetric solution is traceable to a primary standard reference material (SRM) from the National Institute of Standards and Technology, Gaithersburg, USA (NIST SRM 84 potassium hydrogen phthalate) by means of volumetric standard potassium hydrogen phthalate (article number 1.02400), certified reference material according to ISO 17034, analyzed by our accredited calibration laboratory of Merck KGaA, Darmstadt, Germany according to DIN EN ISO/IEC 17025. The uncertainty is expressed as expanded measurement uncertainty with a coverage factor k=2 covering a confidence level of 95%.

Note: The titer is a correction factor to correct for variations of the volumetric solution, the titration equipment, the temperature and other laboratory conditions. For correct titration results it is recommended to determine a titer with the laboratory specific equipment and under laboratory specific conditions directly after opening a new bottle and at regular time intervals.

Legal Information

Titripur is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Met. Corr. 1 - Skin Irrit. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

44.6 °F

Flash Point(C)

7 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service