Skip to Content
Merck
All Photos(1)

Key Documents

1.10887

Supelco

Dichloroisocyanuric acid sodium salt dihydrate

Synonym(s):

Dichloroisocyanuric acid sodium salt dihydrate, 1,3-Dichloro-1,3,5-triazine-2,4,6-trione sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

2.5 KG
€218.00

€218.00


Estimated to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
2.5 KG
€218.00

About This Item

Empirical Formula (Hill Notation):
C3Cl2N3NaO3 · 2H2O
CAS Number:
Molecular Weight:
255.98
MDL number:
UNSPSC Code:
12352107
EC Index Number:
220-767-7
NACRES:
NA.21

€218.00


Estimated to ship on31 March 2025Details


Request a Bulk Order

vapor pressure

<1 hPa ( 20 °C)

Quality Level

form

solid

potency

550-1600 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rabbit)

pH

6.7 (20 °C, 10 g/L in H2O)

mp

252 °C (decomposition)

solubility

236.8 g/L

density

1.97 g/cm3 at 25 °C

bulk density

980 kg/m3

storage temp.

15-25°C

SMILES string

[Na+].ClN1C(=O)[N-]C(=O)N(C1=O)Cl.O.O

InChI

1S/C3HCl2N3O3.Na.2H2O/c4-7-1(9)6-2(10)8(5)3(7)11;;;/h(H,6,9,10);;2*1H2/q;+1;;/p-1

InChI key

PYILKOIEIHHYGD-UHFFFAOYSA-M

Analysis Note

Assay (iodometric, calc. on dried substance): ≥ 98.0 %
Identity (Raman-spectrum): conforms
Loss on Drying (105°C): 9.0 - 16.0 %

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service