Skip to Content
Merck
All Photos(1)

Key Documents

1.10832

Millipore

Geduran® Si 60

for column chromatography (0.063-0.200 mm)

Synonym(s):

Silica, Amorphous Silica, Silicon dioxide

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
€66.00
2.5 KG
€78.00
1108329025
€630.00

€66.00


In StockDetails



Select a Size

Change View
1 KG
€66.00
2.5 KG
€78.00
1108329025
€630.00

About This Item

Linear Formula:
SiO2
CAS Number:
Molecular Weight:
60.08
MDL number:
UNSPSC Code:
41115711
EC Index Number:
231-545-4

€66.00


In StockDetails


Quality Level

potency

>5000 mg/kg LD50, skin (Rabbit)

pH

7 (20 °C, 100 g/L in H2O, slurry)

bp

2230 °C (lit.)

mp

>1600 °C (lit.)

bulk density

200‑800 kg/m3

storage temp.

no temp limit

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

pH-value (10 % suspension): 6.5 - 7.5
Fe (Iron): ≤ 0.02 %
Loss on drying (150 °C): ≤ 7.0 %
Particle size: 90 % between 0.063 - 0.200 mm

Legal Information

GEDURAN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service