Skip to Content
Merck
All Photos(1)

Key Documents

1.10695

Millipore

N-[Tris(hydroxymethyl)-methyl]-2-aminoethanesulfonic acid

Buffer substance TES

Synonym(s):

N-[Tris(hydroxymethyl)-methyl]-2-aminoethanesulfonic acid, TES

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
€342.00
10 KG
€7,970.00

€342.00


Estimated to ship on11 April 2025Details


Request a Bulk Order

Select a Size

Change View
250 G
€342.00
10 KG
€7,970.00

About This Item

CAS Number:
Molecular Weight:
229.24
MDL number:
UNSPSC Code:
12161703
EC Index Number:
230-906-3
NACRES:
NA.05

€342.00


Estimated to ship on11 April 2025Details


Request a Bulk Order

Quality Level

form

solid

mp

226-228 °C

storage temp.

2-30°C

SMILES string

[S](=O)(=O)([O-])CC[N+H2]C(CO)(CO)CO

InChI

1S/C6H15NO6S/c8-3-6(4-9,5-10)7-1-2-14(11,12)13/h7-10H,1-5H2,(H,11,12,13)

InChI key

JOCBASBOOFNAJA-UHFFFAOYSA-N

General description

Buffer substance TES

Application

N-[Tris(hydroxymethyl)-methyl]-2-aminoethanesulfonic acid Buffer substance TES. CAS 7365-44-8, molar mass 229.24 g/mol.

Analysis Note

Assay (alkalimetric)≥ 99 %Identity (IR-spectrum)passes testUV-Absorption (250 nm, 50 g/l, 1 cm, water)≤ 0.05Sulphate (SO₄)≤ 0.005 %Heavy metals (as Pb)≤ 0.0005 %Na (Sodium)≤ 0.1 %Loss on Drying (105°C)≤ 0.2 %Biological buffer, pKa at 20 °C7.50

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service