1.10232
pH-indicator paper pH 1 - 14 Universal indicator
roll L 4.8 m, pH range 1-14, MQuant®
Synonym(s):
pH chemical test paper paper pH 1 - 14 Universal indicator, pH test paper paper pH 1 - 14 Universal indicator
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
product line
MQuant®
Quality Level
feature
non-bleeding: no
refill
packaging
pkg of 6 rolls
roll L
4.8 m
pH range
1-14
detection method
colorimetric
storage temp.
15-25°C
General description
6 replacement rolls (of 4.8 m), for 110962
MQuant® pH indicator papers supplied in rolls are perfectly suited for long-term storage, since they are protected against external influences, such as moisture, light, and ambient gases. These pH indicator papers are manufactured by impregnating high-quality filter paper with indicator or mixed-indicator solutions. The paper is always ready to use; it is easily pulled out of the plastic reel and can be torn off in the necessary length. The pH value is measured semi-quantitatively by visual comparison of the color formed on the paper with the fields of the color scale. This article contains replacement rolls without color scale for article 10962.
MQuant® pH indicator papers supplied in rolls are perfectly suited for long-term storage, since they are protected against external influences, such as moisture, light, and ambient gases. These pH indicator papers are manufactured by impregnating high-quality filter paper with indicator or mixed-indicator solutions. The paper is always ready to use; it is easily pulled out of the plastic reel and can be torn off in the necessary length. The pH value is measured semi-quantitatively by visual comparison of the color formed on the paper with the fields of the color scale. This article contains replacement rolls without color scale for article 10962.
Analysis Note
Application test: conforms
Legal Information
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service