Skip to Content
Merck
All Photos(1)

Key Documents

1.09949

Supelco

Magnesium standard

1000 mg Mg, (MgCl₂ in 6% HCl) Titrisol®

Synonym(s):

Magnesium Reference Standard for ICP/AAS

Sign Into View Organizational & Contract Pricing

Select a Size

1 AMP
€54.90

€54.90


Estimated to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
1 AMP
€54.90

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

€54.90


Estimated to ship on31 March 2025Details


Request a Bulk Order

grade

analytical standard

Quality Level

product line

Titrisol®

form

concentrate

concentration

1000 mg Mg in 6% HCl

technique(s)

AAS: suitable
ICP: suitable

pH

<1 (20 °C in H2O)

density

1.09 g/cm3 at 20 °C

application(s)

industrial qc
pharmaceutical

format

single component solution

storage temp.

15-25°C

Application

Magnesium standard can be used as a calibrationstandard for the determination of metal nutrients in fish feed samples using flameatomic absorption spectrometry (FAAS) [1]

Features and Benefits

  • Titrisol® offers a wide range of concentrated single-element standards.
  • Dilute the concentrated solution to achieve the desired standard concentration.
  • Directly traceable to standard reference materials (SRM) from the National Institute of Standards and Technology (NIST).
  • Supplied in easy-to-handle plastic ampoules.
  • Accompanied with a batch-specific certificate of analysis (COA) containing information on the NIST batch and minimum shelf life.
  • Provides reliable and reproducible results.

Analysis Note

1000 mg Mg in 6% hydrochloric acid, prepared with high-purity MgCl2, HCl, and water.

Legal Information

Titrisol is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service