1.09939
Arsenic standard
1000 mg As, (As₂O₅ in H₂O) Titrisol®
Synonym(s):
Arsenic Standard Solution for ICP/AAS
About This Item
Recommended Products
grade
analytical standard
Quality Level
product line
Titrisol®
form
concentrate
concentration
1000 mg As in H2O
technique(s)
AAS: suitable
ICP: suitable
pH
1.7 (20 °C in H2O)
density
1.02 g/cm3 at 20 °C
application(s)
industrial qc
pharmaceutical
format
single component solution
storage temp.
15-25°C
Related Categories
Application
- Arsenic calibration standard in heavy metal toxicity research: A study on the impact of agricultural practices on heavy metal toxicity, including arsenic, used arsenic calibration standards to assess the levels in food chains, emphasizing the need for precise trace metal analysis in environmental monitoring (Sheema et al., 2024).
- Arsenic standards in environmental health: Editorial discussing the serious arsenic problem highlights the use of arsenic standards in geo-spatial epidemiological studies to identify and address environmental health risks, underscoring the role of high-purity arsenic standards in public health research (Pizzorno, 2024).
- Trace metal analysis using arsenic standards: A study involving the identification of metal(loid) sources in a former mining district utilized arsenic calibration standards for spectroscopy to provide accurate environmental assessments, crucial for managing contaminated sites (Calas et al., 2024).
- Arsenic quantification in epidemiology: Research on the use of arsenic standards to assess environmental exposure levels, illustrating the importance of arsenic quantification standards in epidemiological studies (Sakamoto et al., 2024).
- Arsenic reference material in aquatic studies: Analysis of dissolved inorganic arsenic in the Yellow River used arsenic reference materials to measure seasonal variations and influencing factors, highlighting the application of arsenic standards in aquatic environmental studies (He et al., 2024).
Analysis Note
Determination method: ICP - OES
(traceable of NIST - SRM 3103a)
Accuracy of the method:+/- 5 mg
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Carc. 1A - Eye Dam. 1 - Skin Corr. 1
Storage Class Code
6.1B - Non-combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
EU REACH SVHC Candidate List
EU REACH Annex XVII (Restriction List)
EU REACH Annex XIV (Authorisation List)
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service