Skip to Content
Merck
All Photos(1)

Documents

1.09843

Sigma-Aldrich

Neo-Clear Xylene Substitute

(xylene substitute) for microscopy

Synonym(s):

Neo-Clear

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116113
EC Index Number:
265-067-2
NACRES:
NA.21

vapor pressure

6 hPa ( 20 °C)

Quality Level

form

liquid

autoignition temp.

240 °C

potency

>5000 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rabbit)

expl. lim.

0.6 % (v/v)

IVD

for in vitro diagnostic use

color

clear

refractive index

n20/D 1.418

kinematic viscosity

1.3 cSt(20 °C)

mp

-20 °C

transition temp

flash point 43 °C

density

0.78-0.795 g/cm3 at 20 °C

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

15-25°C

General description

Neo-Clear (xylene substitute) - for microscopy, is an aliphatic hydrocarbon mixture that can substitute the same applications as xylene in the histo-medical lab - histoprocessing, deparaffination and clearing after dehydration in staining process. It is not necessary to change methods or incubation times when using Neo-Clear instead of xylene. As the evaporation rate of Neo-Clear is lower then xylene, it has less odor and the disturbing smell in the lab is reduced significantly. Neo-Clear is more sensitive against water in solvents (turbidity can occur), thus, it is essential to avoid technical qualities of solvents. It is an IVD registered product and CE certified, thus can be used for clinical diagnostic purposes. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Legal Information

NEO-CLEAR is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Asp. Tox. 1 - Flam. Liq. 3

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

110.3 °F

Flash Point(C)

43.5 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Related Content

Learn about the criticality of biological tissue staining for research and clinical pathology using standard and special stains and dyes.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service