Skip to Content
Merck
All Photos(1)

Key Documents

1.09469

Supelco

Multi-element standard II

dissolved in oil 100 ppm: Ag, Al, B, Ba, Ca, Cd, Cr, Cu, Fe, Mg, Mn, Mo, Na, Ni, P, Pb, Si, Sn, Ti, V, Zn Certipur®

Synonym(s):

ICP/AAS Standard for Multiple Elements

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€417.00

€417.00


Estimated to ship on01 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 ML
€417.00

About This Item

UNSPSC Code:
41116107
EC Index Number:
232-384-2
NACRES:
NA.24

€417.00


Estimated to ship on01 April 2025Details


Request a Bulk Order

grade

certified reference material

Quality Level

product line

Certipur®

concentration

100 ppm in oil

technique(s)

AAS: suitable
ICP: suitable

kinematic viscosity

70 cSt

bp

<300 °C

density

0.87 g/cm3 at 20 °C

application(s)

industrial qc
pharmaceutical

format

multi-component solution

storage temp.

15-25°C

General description

This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. This CRM is traceable to SI unit kg and measured against primary material from a National Metrology Institute (NMI), e.g. NIST.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Download your certificate at https://www.sigma-aldrich.com to view certified values, including uncertainty, date of expiry, and detailed information about trace impurities.

Analysis Note

Ag (Silver): 95 – 105
Al (Aluminium): 95 – 105
B (Boron): 95 – 105
Ba (Barium): 95 – 105
Ca (Calcium): 95 – 105
Cd (Cadmium): 95 – 105
Cr (Chromium): 95 – 105
Cu (Copper): 95 – 105
Fe (Iron): 95 – 105
Mg (Magnesium): 95 – 105
Mn (Manganese): 95 – 105
Mo (Molybdenum): 95 – 105
Na (Sodium): 95 – 105
Ni (Nickel): 95 – 105
P (phosphorus): 95 – 105
Pb (Lead): 95 – 105
Si (Silicon): 95 – 105
Sn (Tin): 95 – 105
Ti (Titanium): 95 – 105
V (Vanadium): 95 – 105
Zn (Zinc): 95 – 105
Determination methods: ICP OES, ICP MS, GFAA

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service