Skip to Content
Merck
All Photos(1)

Key Documents

1.09278

Sigma-Aldrich

Wright′s eosine methylene blue

for microscopy

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€104.00
1 KG
€3,860.00

€104.00


Estimated to ship on02 January 2026Details


Request a Bulk Order

Select a Size

Change View
25 G
€104.00
1 KG
€3,860.00

About This Item

UNSPSC Code:
41116130
NACRES:
NA.05

€104.00


Estimated to ship on02 January 2026Details


Request a Bulk Order

Quality Level

form

solid

IVD

for in vitro diagnostic use

technique(s)

microbe id | staining: suitable

pH

6.0-7.0

bulk density

520 kg/m3

εmax

1200-1400 at 648-651 nm in methanol at 0.0005%
650-850 at 521-524 nm in methanol at 0.0005%

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-30°C

General description

Wright′s eosin methylene blue - for microscopy staining dye, is a defined mixture of the dyes C.I. 52015 and C.I. 45380 for hematological staining procedures and is used in conjunction with eosin Y. The Wright′s eosin methylene blue solution can be used for analysis and diagnostics in the whole blood differentiation. The staining of the nuclei is due of the molecular interaction of the Eosin Y dye and a complex of Azur B with DNA. Both dyes assemble to an Eosin Y - Azur B-DNA complex and the intensity of the resulting stain depends on the content of Azur B and the ratio of Azur B : Eosin Y. Furthermore, the resulting stain can vary depending on the influence of fixation, staining times, pH-value of the solutions or buffer substances. For more details, please see instructions for use (IFU). This product is registered as IVD and CE marked. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Analysis Note

Identity (UV/VIS-Spectrum): passes test
pH-value (0,01 %; methanol 50 %): 6.0 - 7.0
Absorption maximum λ 1 (Methanol): 648 - 651 nm
Absorption maximum λ 2 (Methanol): 521 - 524 nm
Spec. Absorptivity A 1%/1cm
- λ1max; 0.0005 %; methanol; calculated on dried substance:
1200 - 1400
- λ2max; 0.0005 %; methanol; calculated on dried substance: 650 - 850
TLC-Test: passes test
Loss on drying (105 °C): ≤ 10 %
Suitability for microscopy: passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service