Skip to Content
Merck
All Photos(1)

Key Documents

1.08541

Supelco

Spray nozzles for TLC sprayer

suitable for thin layer chromatography (TLC)

Sign Into View Organizational & Contract Pricing

Select a Size

6 UNITS
€188.00

€188.00


Available to ship on03 April 2025Details


Request a Bulk Order

Select a Size

Change View
6 UNITS
€188.00

About This Item

UNSPSC Code:
41115711
NACRES:
NB.21

€188.00


Available to ship on03 April 2025Details


Request a Bulk Order

technique(s)

thin layer chromatography (TLC): suitable

storage temp.

2-30°C

General description

Contents: 6 units 5 units 0.8mm, 1 unit 1.25mm
Our TLC accessories allow professionals to work more quickly, more efficiently, and more reliably. Thanks to our broad assortment, users will always find what they need.

Analysis Note

TLC-Test: passes test

related product

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service