Skip to Content
Merck
All Photos(1)

Key Documents

1.08163

Supelco

UV-VIS Standard 3: Sodium iodide solution

according to Ph. Eur. stray light testing, Certipur®

Synonym(s):

UV-VIS Standard 3 - Sodium iodide 10 g/l H2O solution

Sign Into View Organizational & Contract Pricing

Select a Size

3 X 10 ML
€335.00

€335.00


Estimated to ship on03 April 2025Details


Request a Bulk Order

Select a Size

Change View
3 X 10 ML
€335.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

€335.00


Estimated to ship on03 April 2025Details


Request a Bulk Order

grade

analytical standard

Quality Level

Agency

according to Ph. Eur. stray light testing

product line

Certipur®

technique(s)

UV/Vis spectroscopy: suitable

application(s)

environmental
industrial qc

format

mixture

storage temp.

15-25°C

General description

for stray light testing according to Ph Eur Certipur®

Application

UV-VIS Standard 3: Sodium iodide solution as a reference material for checking the stray light spectrophotometers in accordance with European Pharmacopoeia (Ph. Eur.) chapter 2.2.25 and American Society for Testing and Materials (ASTM) E 387 and E 925.

Features and Benefits

  • It is a 10 g/L solution of sodium iodide in H2O
  • Conveniently packed in 10 mL glass ampules. Each box includes 3 single-use ampules
  • Stable solutions in glass ampoules
  • Accreditation according to ISO 17025
  • Traceability to standard reference material from the National Institute of Standards and Technology (NIST) and Physical-Technical Institute (PTB)
  • Includes a certificate of analysis (CoA)
  • Reliable and reproducible results

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service