Skip to Content
Merck
All Photos(1)

Key Documents

1.07534

Supelco

Pyrogallol red

metal indicator

Synonym(s):

Pyrogallol red, Pyrogallic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€201.00

€201.00


Estimated to ship on16 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 G
€201.00

About This Item

Empirical Formula (Hill Notation):
C19H12O8S
CAS Number:
Molecular Weight:
400.36
MDL number:
UNSPSC Code:
12171502
EC Index Number:
251-134-3
NACRES:
NA.21

€201.00


Estimated to ship on16 April 2025Details


Request a Bulk Order

Product Name

Pyrogallol red, metal indicator

Quality Level

form

solid

loss

≤5.0% loss on drying, 110°C

mp

>300 °C

storage temp.

2-30°C

InChI

1S/C19H12O8S/c20-12-7-5-10-15(9-3-1-2-4-14(9)28(24,25)26)11-6-8-13(21)17(23)19(11)27-18(10)16(12)22/h1-8,20,22-23H,(H,24,25,26)/p-1

InChI key

JTZKNOUMMWIARI-UHFFFAOYSA-M

Application


  • Relationship between the structures of flavonoids and oxygen radical absorbance capacity values: a quantum chemical analysis.: This study explores the role of Pyrogallol Red in analyzing the antioxidant properties of flavonoids, providing insights into its utility in quantifying oxygen radical absorbance capacities, essential for evaluating the antioxidative potential of various substances in analytical chemistry applications (Zhang et al., 2013).


Analysis Note

Appearance of solution: conforms
Loss on drying (110 °C): ≤ 5.0 %
Suitability as indicator (for metal titration): conforms

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service