Skip to Content
Merck

1.07133

SAFC

Pancreatin

1400 FIP-U/g Protease, 24000 FIP-U/g Lipase, 30000 FIP-U/g Amylase EMPROVE® ESSENTIAL Ph Eur,4 x USP

Pharma Manufacturing

Synonym(s):

Pancreatin

Sign Into View Organizational & Contract Pricing

Select a Size

5 KG
€1,460.00

€1,460.00


Estimated to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
5 KG
€1,460.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352204
EC Index Number:
232-468-9

€1,460.00


Estimated to ship on22 April 2025Details


Request a Bulk Order

Agency

Ph. Eur.
USP

Quality Level

product line

EMPROVE® ESSENTIAL

form

solid

autoignition temp.

>360 °C

potency

>10000 mg/kg LD50, oral (Rat)

pH

5-6 (20 °C, 10 g/L in H2O)

bulk density

400 kg/m3

application(s)

pharmaceutical
solid formulation

storage temp.

2-8°C

General description

We offer a range of high quality critical raw materials, solvents and reagents plus the documentation and expertise to meet stringent regulatory requirements. In addition to our comprehensive portfolio, we also offer custom manufacturing services to meet your exact needs. As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Pancreatin is a digestive enzyme used to supplement the body′s natural supply of such enzymes. Pancreatin Emprove® Essential is suitable for pharmaceutical applications.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service