1.06446
tri-Sodium citrate dihydrate
powder, EMPROVE® ESSENTIAL, Ph. Eur., BP, JP, USP, E 331
Pharma Manufacturing
Synonym(s):
Sodium citrate tribasic dihydrate, Trisodium Citrate, Sodium Citrate, Dihydrate, Citric acid trisodium salt dihydrate, Trisodium citrate dihydrate
About This Item
Recommended Products
Agency
BP
JP
Ph. Eur.
USP
Quality Level
product line
EMPROVE® ESSENTIAL
form
powder
solid
pH
7.5-9.0 (25 °C, 50 g/L in H2O)
mp
150 °C (anhydrous substance)
>300 °C (lit.)
application(s)
liquid formulation
ophthalmics
pharmaceutical
semi-solid formulation
solid formulation
storage temp.
2-30°C
SMILES string
O.O.[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
InChI
1S/C6H8O7.3Na.2H2O/c7-3(8)1-6(13,5(11)12)2-4(9)10;;;;;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);;;;2*1H2/q;3*+1;;/p-3
InChI key
NLJMYIDDQXHKNR-UHFFFAOYSA-K
Looking for similar products? Visit Product Comparison Guide
General description
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.
Application
Legal Information
also commonly purchased with this product
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service