Skip to Content
Merck
All Photos(1)

Key Documents

1.06246

Supelco

1-Naphtholphthalein

pH indicator

Synonym(s):

1-Naphtholphthalein

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€358.00
5 G
€409.00
100 G
€3,580.00

€358.00


Estimated to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
1 G
€358.00
5 G
€409.00
100 G
€3,580.00

About This Item

Empirical Formula (Hill Notation):
C28H18O4
CAS Number:
Molecular Weight:
418.44
MDL number:
UNSPSC Code:
12352115
EC Index Number:
209-875-5
NACRES:
NA.21

€358.00


Estimated to ship on31 March 2025Details


Request a Bulk Order

Product Name

1-Naphtholphthalein, indicator

Quality Level

form

solid

loss

≤5% loss on drying, 110°C

visual transition interval

7.1-8.3, pink-brown to blue

mp

253-255 °C

bulk density

350 kg/m3

Amax

470-485 nm (buffer pH 7.1)
649-654 nm (buffer pH 8.3)

storage temp.

2-30°C

SMILES string

O1C(c6c(cccc6)C1=O)(c4c5c(c(cc4)O)cccc5)c2c3c(c(cc2)O)cccc3

InChI

1S/C28H18O4/c29-25-15-13-23(17-7-1-3-9-19(17)25)28(22-12-6-5-11-21(22)27(31)32-28)24-14-16-26(30)20-10-4-2-8-18(20)24/h1-16,29-30H

InChI key

HQHBAGKIEAOSNM-UHFFFAOYSA-N

Analysis Note

Identity (UV/VIS-Spectrum): passes test
Transition range: pH 7.1 - pH 8.3pinkbrownish - blue
Absorption maximum λ 1 (buffer pH 7.1): 470 - 485 nm
Absorption maximum λ 2 (buffer pH 8.3): 649 - 654 nm
Spec. Absorptivity A 1%/1cm (λ1max; 0.01 g/l; buffer pH 7.1; calc. on anhydrous substance): 100 - 180
Spec. Absorptivity A 1%/1cm (λ2max; 0.01 g/l; buffer pH 8.3; calc. on anhydrous substance): 360 - 520
Loss on drying (110 °C): ≤ 5 %

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service