Skip to Content
Merck
All Photos(1)

Key Documents

1.06130

Supelco

Mixed indicator 5

pH indicator, for ammonia titrations

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
€71.50
1 L
€223.00

€71.50


Estimated to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
250 ML
€71.50
1 L
€223.00

About This Item

UNSPSC Code:
12352401
NACRES:
NB.21

€71.50


Estimated to ship on31 March 2025Details


Request a Bulk Order

Product Name

Mixed indicator 5, for ammonia titrations

Quality Level

autoignition temp.

425 °C (ethanol)

pH

4.4-5.8 (20 °C in H2O)

transition temp

flash point 14 °C

density

0.82 g/cm3 at 20 °C

storage temp.

15-25°C

Analysis Note

Application test: conforms
pH-value (20 °C): 5.0 - 5.5

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F

Flash Point(C)

14 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service