Skip to Content
Merck
All Photos(1)

Key Documents

1.06126

Millipore

2-Morpholinoethanesulfonic acid monohydrate

buffer substance MES

Synonym(s):

2-Morpholinoethanesulfonic acid monohydrate, MES

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€50.80
250 G
€171.00
10 KG
€3,560.00

€50.80


Estimated to ship on07 April 2025Details


Request a Bulk Order

Select a Size

Change View
25 G
€50.80
250 G
€171.00
10 KG
€3,560.00

About This Item

CAS Number:
Molecular Weight:
213.25
MDL number:
UNSPSC Code:
12161703
EC Index Number:
224-632-3
NACRES:
NA.21

€50.80


Estimated to ship on07 April 2025Details


Request a Bulk Order

Quality Level

form

powder

pH

3.6 (20 °C, 10 g/L in H2O)

mp

>300 °C

solubility

80 g/L

bulk density

570 kg/m3

storage temp.

2-30°C

SMILES string

[S](=O)(=O)(O)CCN1CCOCC1

InChI

1S/C6H13NO4S/c8-12(9,10)6-3-7-1-4-11-5-2-7/h1-6H2,(H,8,9,10)

InChI key

SXGZJKUKBWWHRA-UHFFFAOYSA-N

General description

buffer substance MES

Application

2-Morpholinoethanesulfonic acid monohydrate buffer substance MES. CAS 145224-94-8, pH 3.6 (10 g/l, H₂O, 20 °C).

Analysis Note

Assay (alcalimetric, calculated on anhydrous substance)≥ 99 %Identity (IR-spectrum)passes testUV-Absorption (250 nm, 50 g/l, 1 cm, water)≤ 0.05Chloride (Cl)≤ 0.005 %Sulphate (SO₄)≤ 0.005 %Heavy metals (as Pb)≤ 0.0005 %Na (Sodium)≤ 0.005 %Water (according to Karl Fischer)7.0 - 9.0 %Biological buffer, pKa at 20 °C6.15

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service