Skip to Content
Merck
All Photos(1)

Key Documents

1.05912

Millipore

Maltose monohydrate

for biochemistry

Synonym(s):

Maltose monohydrate

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€74.60
100 G
€256.00

€74.60


Estimated to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
25 G
€74.60
100 G
€256.00

About This Item

CAS Number:
Molecular Weight:
360.32
MDL number:
UNSPSC Code:
12352201
EC Index Number:
200-716-5
NACRES:
NA.79

€74.60


Estimated to ship on31 March 2025Details


Request a Bulk Order

Quality Level

form

solid

pH

4.5-6.0 (20 °C, 50 g/L in H2O)

mp

125 °C (anhydrous)

solubility

470.2 g/L

bulk density

320 kg/m3

storage temp.

15-25°C

SMILES string

O1[C@@H]([C@@H]([C@H]([C@@H]([C@H]1CO)O)O)O)O[C@@H]([C@H](O)[C@@H](O)C=O)[C@H](O)CO.O

InChI

1S/C12H22O11.H2O/c13-1-4(16)7(18)11(5(17)2-14)23-12-10(21)9(20)8(19)6(3-15)22-12;/h1,4-12,14-21H,2-3H2;1H2/t4-,5+,6+,7+,8+,9-,10+,11+,12+;/m0./s1

InChI key

HBDJFVFTHLOSDW-DNDLZOGFSA-N

General description

for biochemistry

Application

Maltose monohydrate for biochemistry. CAS 6363-53-7, pH 4.5 - 6.0 (50 g/l, H₂O, 20 °C).

Analysis Note

Purity (HPLC)≥ 98.0 %Identity (IR)conformsSpec. rotation [α²0/D (10 %; water; calc. on anhydrous substance)+135 - +139 °N (Nitrogen)≤ 0.1 %Heavy metals (as Pb)≤ 0.001 %Fe (Iron)≤ 0.0005 %Methanol (HS-GC)≤ 3000 ppmResidual solvents class 3(Ph Eur/ICH)≤ 0.5 %Other residual solvents (Ph Eur/ICH)excluded by production processGlucose (HPLC)≤ 0.5 %Sulfated ash≤ 0.1 %Water5.0 - 5.5 %

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service