Skip to Content
Merck
All Photos(1)

Key Documents

1.05829

SAFC

Magnesium hydroxide carbonate

heavy extra pure, Ph. Eur., BP, USP, E 504

Pharma Manufacturing

Synonym(s):

Magnesium hydroxide carbonate, Magnesium carbonate basic

Sign Into View Organizational & Contract Pricing

Select a Size

40 KG
€630.00

€630.00


Estimated to ship on24 March 2025Details


Request a Bulk Order

Select a Size

Change View
40 KG
€630.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352302
E Number:
E504
EC Index Number:
235-192-7

€630.00


Estimated to ship on24 March 2025Details


Request a Bulk Order

Agency

BP
Ph. Eur.
USP

Quality Level

Assay

40.0-43.5% Mg (as MgO) basis (USP)
40.0-43.5% Mg (as MgO) basis (complexometric)

form

solid

pH

10.5 (20 °C, 50 g/L in H2O, suspension)

mp

600 °C (decomposition)

solubility

water: insoluble at 20 °C

density

2.16 g/cm3 at 20 °C

anion traces

chloride (Cl-): ≤0.02%
sulfate (SO42-): ≤0.3%

cation traces

As: ≤0.0002%
Ca: ≤0.4%
Fe: ≤0.005%
Hg: ≤0.0001%
Na: ≤0.1%
Ni: ≤0.0100%
Pb: ≤0.0002%
Si: ≤0.05%
V: ≤0.0010%
heavy metals (as Pb): ≤0.001%

application(s)

pharma/biopharma processes

storage temp.

2-30°C

SMILES string

[Mg+2].[O-]C(=O)[O-].[O-H]

InChI

1S/CH2O3.Mg.H2O/c2-1(3)4;;/h(H2,2,3,4);;1H2/q;+2;/p-3

InChI key

OUHCLAKJJGMPSW-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

We offer a range of high quality critical raw materials plus the documentation and expertise to help our customers meet stringent regulatory requirements. Our SAFC® portfolio of high-quality products withstands strict quality control procedures.

Application

Magnesium hydroxide carbonate can be used to improve flow properties of dry powders in pharmaceutical processes.

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service