Skip to Content
Merck
All Photos(1)

Documents

1.05813

Supelco

Magnesium matrix modifier

for graphite furnace AAS c(Mg) = 10.0 ± 0.2 g/l (Mg(NO₃)₂ * 6 H₂O in HNO₃ ca. 17%)

Synonym(s):

Magnesium Enhancement Solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.21

grade

for inorganic trace analysis

Quality Level

technique(s)

AAS: suitable

pH

<1 (20 °C in H2O)

density

1.05 g/cm3 at 20 °C

application(s)

general analytical

storage temp.

2-30°C

Analysis Note

Ag (Silver): ≤ 0.000002 g/l
As (Arsenic): ≤ 0.00001 g/l
Cd (Cadmium): ≤ 0.000001 g/l
Pb (Lead): ≤ 0.00001 g/l
Se (Selenium): ≤ 0.0001 g/l
Tl (Thallium): ≤ 0.00001 g/l

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service